5-Chloro-2-cyano-3-nitropyridine

Lieferant: Apollo Scientific
Gefahr

Synonyme: 5-Chloro-3-nitropyridine-2-carbonitrile

OR18600-100G OR18600-25G
APOSOR18600-100GEA 260 EUR
APOSOR18600-100G APOSOR18600-25G
5-Chloro-2-cyano-3-nitropyridine
5-Chloro-2-cyano-3-nitropyridine
Formel: C6H2ClN3O2
Lagertemperatur: Raumtemperatur
MDL: MFCD06657552
CAS-Nummer: 181123-11-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR