Oxypurinol 95%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4,6-Dihydroxypyrazolo[3,4-d]pyrimidine, 1H-Pyrazolo[3,4-d]pyrimidine-4,6-diol

OR18615-1G OR18615-5G OR18615-250MG
APOSOR18615-1GEA 59.6 EUR
APOSOR18615-1G APOSOR18615-5G APOSOR18615-250MG
Oxypurinol 95%
Oxypurinol
Formel: C₅H₄N₄O₂
Molecular Weight: 152,11 g/mol
Schmelzpunkt: >300 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00047205
CAS-Nummer: 2465-59-0
EINECS: 219-570-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR