3-tert-Butylisoxazole-5-amine
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
3-(tert-Butyl)isoxazol-5-amine
OR18617-1G
OR18617-5G
OR18617-10G
APOSOR18617-1GEA
23.2
EUR
APOSOR18617-1G
APOSOR18617-5G
APOSOR18617-10G
3-tert-Butylisoxazole-5-amine
3-tert-Butylisoxazole-5-amine
Formel:
C₇H₁₂N₂O Molecular Weight: 140,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD05266697 CAS-Nummer: 59669-59-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...