2,5-Dihydroxybenzaldehyd 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Gentisaldehyd

OR1868-25G OR1868-100G OR1868-10G
APOSOR1868-25GEA 72.5 EUR
APOSOR1868-25G APOSOR1868-100G APOSOR1868-10G
2,5-Dihydroxybenzaldehyd 98%
2,5-Dihydroxybenzaldehyd
Formel: C₇H₆O₃
Molecular Weight: 138.12 g/mol
Schmelzpunkt: 97…103 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00003333
CAS-Nummer: 1194-98-5
EINECS: 214-789-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR