3-Azetidinolhydrochlorid 97%
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
3-Hydroxyazetidine hydrochloride
OR1873-100G
OR1873-25G
APOSOR1873-100GEA
63.8
EUR
APOSOR1873-100G
APOSOR1873-25G
3-Azetidinolhydrochlorid 97%
Azetidin-3-ol hydrochloride
Formel:
C₃H₈ClNO Molecular Weight: 109,56 g/mol Schmelzpunkt: 85…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02683887 CAS-Nummer: 18621-18-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...