(3-Iodphenyl)essigsäure 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 2-(3-Iodophenyl)acetic acid

OR18733-25G OR18733-5G OR18733-1G
APOSOR18733-25GEA 178 EUR
APOSOR18733-25G APOSOR18733-5G APOSOR18733-1G
(3-Iodphenyl)essigsäure 97%
(3-Iodphenyl)essigsäure
Formel: (C₆H₄I)CH₂COOH
Molecular Weight: 262.05 g/mol
Schmelzpunkt: 127…131 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00046548
CAS-Nummer: 1878-69-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR