2,3,5-Tribrom-4-methylthiophen

Lieferant: Apollo Scientific

Synonyme: 3-Methyl-2,4,5-tribromothiophene

OR18820-10G OR18820-5G
APOSOR18820-10GEA 46.4 EUR
APOSOR18820-10G APOSOR18820-5G
2,3,5-Tribrom-4-methylthiophen
2,3,5-Tribromo-4-methylthiophene
Formel: C₅H₃Br₃S
Molecular Weight: 334,85 g/mol
Schmelzpunkt: 31 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00130086
CAS-Nummer: 67869-13-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR