Methyl 4-Formyl-3-nitrobenzoate 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Formyl-3-nitrobenzoic acid methyl ester
OR18823-25G
APOSOR18823-25GEA
79.9
EUR
APOSOR18823-25G
Methyl 4-Formyl-3-nitrobenzoate 98%
Methyl 4-Formyl-3-nitrobenzoate
Formel:
C₉H₇NO₅ Molecular Weight: 209,16 g/mol Schmelzpunkt: 62…66 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04039968 CAS-Nummer: 153813-69-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...