Methyl-5-methylpicolinat 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: Methyl 5-methylpyridine-2-carboxylate, Methyl 5-methylpicolinate

OR18831-5G
APOSOR18831-5GEA 81.2 EUR
APOSOR18831-5G
Methyl-5-methylpicolinat 98%
Methyl-5-methylpicolinat
Formel: C₈H₉NO₂
Molecular Weight: 151,1626 g/mol
Schmelzpunkt: 54…55 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11040648
CAS-Nummer: 29681-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR