1-(3,5-Dichlorophenyl)-1,2-propandione 95+%
Lieferant: Apollo Scientific
OR1885-1G
OR1885-5G
OR1885-25G
APOSOR1885-1GEA
136
EUR
APOSOR1885-1G
APOSOR1885-5G
APOSOR1885-25G
1-(3,5-Dichlorophenyl)-1,2-propandione 95+%
1-(3,5-Dichlorophenyl)-1,2-propandione
Formel:
C₉H₆Cl₂O₂ Schmelzpunkt: 45…46 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01941316 CAS-Nummer: 386715-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...