3-Brom-4-methoxy-5-methylanilin

Lieferant: Apollo Scientific
Achtung

OR18850-1G OR18850-5G OR18850-250MG
APOSOR18850-1GEA 29 EUR
APOSOR18850-1G APOSOR18850-5G APOSOR18850-250MG
3-Brom-4-methoxy-5-methylanilin
3-Brom-4-methoxy-5-methylanilin
Formel: C₈H₁₀BrNO
Lagertemperatur: Raumtemperatur
MDL: MFCD16618926
CAS-Nummer: 1215205-12-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR