6-Brompicolinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
6-Bromo-2-pyridinecarboxylic acid, 6-Bromopyridine-2-carboxylic acid
OR1906-25G
OR1906-5G
OR1906-100G
APOSOR1906-25GEA
36.4
EUR
APOSOR1906-25G
APOSOR1906-5G
APOSOR1906-100G
6-Brompicolinsäure
6-Brompicolinsäure
Formel:
C₆H₄BrNO₂ Molecular Weight: 202.01 g/mol Schmelzpunkt: 188…194 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00093197 CAS-Nummer: 21190-87-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...