5-Aminonicotinsäure
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Aminopyridine-3-carboxylic acid, 5-Amino-3-pyridinecarboxylic acid
OR1915-100G
OR1915-25G
OR1915-5G
APOSOR1915-100GEA
115
EUR
APOSOR1915-100G
APOSOR1915-25G
APOSOR1915-5G
5-Aminonicotinsäure
5-Aminonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: 293 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00129116 CAS-Nummer: 24242-19-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...