3,5-Dimethyl-4-ethoxyphenylboronsäure 98%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Ethoxy-3,5-dimethylbenzeneboronic acid, 4-Ethoxy-3,5-dimethylphenylboronic acid, (4-Ethoxy-3,5-dimethylphenyl)boronic acid, 3,5-Dimethyl-4-ethoxybenzeneboronic acid

OR1931-25G OR1931-5G
APOSOR1931-25GEA 231 EUR
APOSOR1931-25G APOSOR1931-5G
3,5-Dimethyl-4-ethoxyphenylboronsäure 98%
3,5-Dimethyl-4-ethoxyphenylboronsäure
Formel: C₁₀H₁₅BO₃
Schmelzpunkt: 206…212 °C
Lagertemperatur: Kühlschrank
MDL: MFCD02179463
CAS-Nummer: 850568-59-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR