3-Chlor-(4-methoxycarbonyl)phenylboronsäure 96%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
(3-Chloro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(methoxycarbonyl)phenylboronic acid, 3-Chloro-4-(methoxycarbonyl)benzeneboronic acid
OR1933-1G
OR1933-25G
OR1933-5G
APOSOR1933-1GEA
21.9
EUR
APOSOR1933-1G
APOSOR1933-25G
APOSOR1933-5G
3-Chlor-(4-methoxycarbonyl)phenylboronsäure 96%
3-Chlor-(4-methoxycarbonyl)phenylboronsäure
Formel:
C₈H₈BClO₄ Molecular Weight: 214,4100 g/mol Schmelzpunkt: 174…178 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06659874 CAS-Nummer: 603122-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...