2-Bromo-N,N-dimethylpyridin-4-amine 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Bromo-4-(dimethylamino)pyridine
OR19500-1G
OR19500-5G
APOSOR19500-1GEA
58
EUR
APOSOR19500-1G
APOSOR19500-5G
2-Bromo-N,N-dimethylpyridin-4-amine 98%
2-Bromo-N,N-dimethylpyridin-4-amine
Formel:
C₇H₉BrN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD11036210 CAS-Nummer: 396092-82-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...