2,7-Dibromnaphthalin 98%

Lieferant: Apollo Scientific
Achtung

OR19537-10G OR19537-5G OR19537-25G
APOSOR19537-10GEA 60.9 EUR
APOSOR19537-10G APOSOR19537-5G APOSOR19537-25G
2,7-Dibromnaphthalin 98%
2,7-Dibromonaphthalene
Formel: C₁₀H₆Br₂
Molecular Weight: 285.97 g/mol
Schmelzpunkt: 139…142 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00238580
CAS-Nummer: 58556-75-5
UN: 3077
ADR: 9+EHS III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR