2,2'-[ethylenebis(oxy)]bisacetic acid

Lieferant: Apollo Scientific
Achtung

Synonyme: 3,6-Dioxaoctanedioic acid

OR19555-1G OR19555-5G
APOSOR19555-1GEA 43.5 EUR
APOSOR19555-1G APOSOR19555-5G
2,2'-[ethylenebis(oxy)]bisacetic acid
2,2'-[ethylenebis(oxy)]bisacetic acid
Formel: C₆H₁₀O₆
Molecular Weight: 178,14 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00054502
CAS-Nummer: 23243-68-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR