3-Amino-4-phenyl-1H-pyrazole
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-Phenyl-1H-pyrazol-3-amine
OR1969-5G
APOSOR1969-5GEA
89.9
EUR
APOSOR1969-5G
3-Amino-4-phenyl-1H-pyrazole
3-Amino-4-phenyl-1H-pyrazole
Formel:
C₉H₉N₃ Molecular Weight: 159,19 g/mol Schmelzpunkt: 174…177 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01693715 CAS-Nummer: 5591-70-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...