2-(Thiazol-2-yl)benzaldehyde

Lieferant: Apollo Scientific
Gefahr

OR1995-1G OR1995-5G
APOSOR1995-1GEA 108 EUR
APOSOR1995-1G APOSOR1995-5G
2-(Thiazol-2-yl)benzaldehyde
2-(Thiazol-2-yl)benzaldehyde
Formel: C₁₀H₇NOS
Molecular Weight: 189,2300 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD06657778
CAS-Nummer: 223575-69-7
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR