1-[2-(2-Amino-3-methoxyphenyl)ethyl]pyrrolidine 95+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methoxy-6-[2-(pyrrolidin-1-yl)ethyl]aniline
OR1996-250MG
OR1996-1G
APOSOR1996-250MGEA
145
EUR
APOSOR1996-250MG
APOSOR1996-1G
1-[2-(2-Amino-3-methoxyphenyl)ethyl]pyrrolidine 95+%
1-[2-(2-Amino-3-methoxyphenyl)ethyl]pyrrolidine
Formel:
C₁₃H₂₀N₂O Schmelzpunkt: 83…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00202972 CAS-Nummer: 387358-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...