2,5-Dichloro-1H-benzimidazole

Lieferant: Apollo Scientific
Achtung

Synonyme: 2,6-Dichloro-1H-benzo[d]imidazole

OR200016-1G OR200016-250MG OR200016-500MG
APOSOR200016-1GEA 517 EUR
APOSOR200016-1G APOSOR200016-250MG APOSOR200016-500MG
2,5-Dichloro-1H-benzimidazole
2,5-Dichloro-1H-benzimidazole
Formel: C₇H₄Cl₂N₂
Molecular Weight: 187,03 g/mol
Schmelzpunkt: 207…208 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00091850
CAS-Nummer: 4887-95-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR