1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid

Lieferant: Apollo Scientific

Synonyme: 1-[4-(Methylsulphonyl)phenyl]-5-oxo-3-pyrrolidinecarboxylic acid

OR200035-1G OR200035-5G OR200035-10G
APOSOR200035-1GEA 116 EUR
APOSOR200035-1G APOSOR200035-5G APOSOR200035-10G
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid
1-[4-(Methylthio)phenyl]-5-oxopyrrolidine-3-carboxylic acid
Formel: C₁₂H₁₃NO₃S
Schmelzpunkt: 173…175 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08444946
CAS-Nummer: 63674-53-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR