4-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Methyl-1H-benzo[d]imidazol-2(3H)-one

OR200060-1G OR200060-5G
APOSOR200060-1GEA 82.8 EUR
APOSOR200060-1G APOSOR200060-5G
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one 97%
4-Methyl-1,3-dihydro-2H-benzimidazol-2-one
Formel: C₈H₈N₂O
Molecular Weight: 148,17 g/mol
Schmelzpunkt: 273…280 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08274669
CAS-Nummer: 19190-68-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR