4-Methyl-1,3-benzoxazol-2(3H)-one

Lieferant: Apollo Scientific

OR200061-1G OR200061-250MG OR200061-50MG
APOSOR200061-1GEA 491 EUR
APOSOR200061-1G APOSOR200061-250MG APOSOR200061-50MG
4-Methyl-1,3-benzoxazol-2(3H)-one
4-Methyl-1,3-benzoxazol-2(3H)-one
Formel: C₈H₇NO₂
Molecular Weight: 149.15 g/mol
Schmelzpunkt: 153…154 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16742805
CAS-Nummer: 78258-80-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR