N1-Methyl-4-nitro-1,2-phenylenediamine

Lieferant: Apollo Scientific
Achtung

Synonyme: 1,2-Diamino-N1-methyl-4-nitrobenzene, N1-Methyl-4-nitrobenzene-1,2-diamine, 2-Amino-1-methylamino-4-nitrobenzene

OR200062-5G
APOSOR200062-5GEA 29 EUR
APOSOR200062-5G
N1-Methyl-4-nitro-1,2-phenylenediamine
N1-Methyl-4-nitro-1,2-phenylenediamine
Formel: C₇H₉N₃O₂
Molecular Weight: 167,17 g/mol
Schmelzpunkt: 168…179 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00156607
CAS-Nummer: 41939-61-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR