4-Pyrimidinecarboxylic acid, methyl ester
Lieferant: Apollo Scientific
|
Gefahr
|
Synonyme:
Methyl pyrimidine-4-carboxylate
OR200069-1G
OR200069-5G
APOSOR200069-1GEA
36.4
EUR
APOSOR200069-1G
APOSOR200069-5G
4-Pyrimidinecarboxylic acid, methyl ester
4-Pyrimidinecarboxylic acid, methyl ester
Formel:
C₆H₆N₂O₂ Molecular Weight: 138.13 g/mol Schmelzpunkt: 63…71 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234184 CAS-Nummer: 2450-08-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...