Ethyl-3-hydroxy-4-nitrobenzoate
Lieferant: Apollo Scientific
|
Achtung
|
OR200073-1G
OR200073-5G
APOSOR200073-1GEA
69.6
EUR
APOSOR200073-1G
APOSOR200073-5G
Ethyl-3-hydroxy-4-nitrobenzoate
Ethyl-3-hydroxy-4-nitrobenzoate
Formel:
C₉H₉NO₅ Molecular Weight: 211.18 g/mol Schmelzpunkt: 89…90 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06204637 CAS-Nummer: 717-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...