2-Pyridinylessigsäurehydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-(Pyridin-2-yl)acetic acid hydrochloride, 2-(Pyridin-2-yl)acetic acid HCl
OR20009-25G
OR20009-5G
OR20009-100G
APOSOR20009-25GEA
84.1
EUR
APOSOR20009-25G
APOSOR20009-5G
APOSOR20009-100G
2-Pyridinylessigsäurehydrochlorid
2-Pyridylacetic acid hydrochloride
Formel:
C₇H₇NO₂ Molecular Weight: 173.60 g/mol Schmelzpunkt: 135 °C (Zers.) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00012812 CAS-Nummer: 16179-97-8 EINECS: 240-316-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...