Methyl-4-sulphamoylbenzoate

Lieferant: Apollo Scientific

Synonyme: Methyl 4-sulphamoylbenzoate

OR200095-1G OR200095-100MG OR200095-250MG OR200095-500MG OR200095-2.5G
APOSOR200095-1GEA 519 EUR
APOSOR200095-1G APOSOR200095-100MG APOSOR200095-250MG APOSOR200095-500MG APOSOR200095-2.5G
Methyl-4-sulphamoylbenzoate
Methyl-4-sulphamoylbenzoate
Formel: C₈H₉NO₄S
Molecular Weight: 215,23 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03768500
CAS-Nummer: 22808-73-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR