1-[2-(3-Bromophenoxy)ethyl]piperidine

Lieferant: Apollo Scientific

OR200098-1G OR200098-100MG OR200098-250MG OR200098-500MG OR200098-2.5G
APOSOR200098-1GEA 249 EUR
APOSOR200098-1G APOSOR200098-100MG APOSOR200098-250MG APOSOR200098-500MG APOSOR200098-2.5G
1-[2-(3-Bromophenoxy)ethyl]piperidine
1-[2-(3-Bromophenoxy)ethyl]piperidine
Formel: C₁₃H₁₈BrNO
Molecular Weight: 284.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD03992861
CAS-Nummer: 554430-68-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR