1-[2-(2-Bromophenoxy)ethyl]piperidine

Lieferant: Apollo Scientific

OR200099-1G OR200099-250MG OR200099-500MG OR200099-2.5G
APOSOR200099-1GEA 249 EUR
APOSOR200099-1G APOSOR200099-250MG APOSOR200099-500MG APOSOR200099-2.5G
1-[2-(2-Bromophenoxy)ethyl]piperidine
1-[2-(2-Bromophenoxy)ethyl]piperidine
Formel: C₁₃H₁₈BrNO
Molecular Weight: 284.2 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01308393
CAS-Nummer: 864424-01-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR