4-(4-Bromophenoxy)benzonitrile
Lieferant: Apollo Scientific
OR200101-1G
OR200101-250MG
OR200101-500MG
OR200101-2.5G
APOSOR200101-1GEA
310
EUR
APOSOR200101-1G
APOSOR200101-250MG
APOSOR200101-500MG
APOSOR200101-2.5G
4-(4-Bromophenoxy)benzonitrile
4-(4-Bromophenoxy)benzonitrile
Formel:
C₁₃H₈BrNO Lagertemperatur: Raumtemperatur |
MDL:
MFCD08446949 CAS-Nummer: 330792-93-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...