6-Chloro-N-hydroxynicotinamide

Lieferant: Apollo Scientific

OR200102-1G OR200102-250MG OR200102-500MG OR200102-2.5G
APOSOR200102-1GEA 458 EUR
APOSOR200102-1G APOSOR200102-250MG APOSOR200102-500MG APOSOR200102-2.5G
6-Chloro-N-hydroxynicotinamide
6-Chloro-N-hydroxynicotinamide
Formel: C₆H₅ClN₂O₂
Molecular Weight: 172.57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD15144734
CAS-Nummer: 1263094-64-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR