3-Bromo-1-ethyl-1H-pyrazole
Lieferant: Apollo Scientific
OR200118-50MG
OR200118-250MG
APOSOR200118-50MGEA
197
EUR
APOSOR200118-50MG
APOSOR200118-250MG
3-Bromo-1-ethyl-1H-pyrazole
3-Bromo-1-ethyl-1H-pyrazole
Formel:
C₅H₇BrN₂ Lagertemperatur: Raumtemperatur |
MDL:
MFCD09414729 CAS-Nummer: 1216504-91-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...