Methyl-2-(chlorocarbonyl)benzoate

Lieferant: Apollo Scientific
Gefahr

OR200131-1G OR200131-5G
APOSOR200131-1GEA 0 EUR
APOSOR200131-1G APOSOR200131-5G
Methyl-2-(chlorocarbonyl)benzoate
Methyl-2-(chlorocarbonyl)benzoate
Formel: C₉H₇ClO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11656039
CAS-Nummer: 4397-55-1
UN: 3265
ADR: 8,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR