3-(Acetylamino)benzoyl chloride

Lieferant: Apollo Scientific

OR200167-1G OR200167-250MG
APOSOR200167-1GEA 0 EUR
APOSOR200167-1G APOSOR200167-250MG
3-(Acetylamino)benzoyl chloride
3-(Acetylamino)benzoyl chloride
Formel: C₉H₈ClNO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD11939183
CAS-Nummer: 90531-74-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR