4-(Aminomethyl)benzonitril Hydrochlorid
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
4-(Aminomethyl)benzonitrile HCl, α-Amino-p-tolunitrile Hydrochloride, 4-(Aminomethyl)benzonitrile Hydrochloride, 4-Cyanobenzylamine Hydrochloride
OR20018-100G
OR20018-25G
OR20018-500G
APOSOR20018-100GEA
58
EUR
APOSOR20018-100G
APOSOR20018-25G
APOSOR20018-500G
4-(Aminomethyl)benzonitril Hydrochlorid
4-(Aminomethyl)benzonitril Hydrochlorid
Formel:
C₈H₉ClN₂ Schmelzpunkt: 274…279 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD01861472 CAS-Nummer: 15996-76-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...