4-Brom-2-chlorbenzaldehyd 98%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Chloro-4-bromobenzaldehyde
OR2003-100G
OR2003-25G
OR2003-5G
APOSOR2003-100GEA
84.1
EUR
APOSOR2003-100G
APOSOR2003-25G
APOSOR2003-5G
4-Brom-2-chlorbenzaldehyd 98%
4-Bromo-2-chlorobenzaldehyde
Formel:
C₇H₄BrClO Molecular Weight: 219,46 g/mol Schmelzpunkt: 73…75 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08741413 CAS-Nummer: 158435-41-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...