(2-Methyl-3-nitrophenyl)essigsäure 99+%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
2-Methyl-3-nitrophenylacetic Acid
OR2005-100G
OR2005-25G
APOSOR2005-100GEA
111
EUR
APOSOR2005-100G
APOSOR2005-25G
(2-Methyl-3-nitrophenyl)essigsäure 99+%
(2-Methyl-3-nitrophenyl)essigsäure
Formel:
C₉H₉NO₄ Molecular Weight: 195,17 g/mol Schmelzpunkt: 132…136 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD07782132 CAS-Nummer: 23876-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...