2-Ethyl-6-methyl-3-pyridinol
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Emoxypine, 2-Ethyl-6-methyl-3-pyridinol
OR2008-100G
OR2008-10G
OR2008-5G
APOSOR2008-100GEA
130
EUR
APOSOR2008-100G
APOSOR2008-10G
APOSOR2008-5G
2-Ethyl-6-methyl-3-pyridinol
2-Ethyl-3-hydroxy-6-methylpyridine
Formel:
C₈H₁₁NO Molecular Weight: 137,1800 g/mol Schmelzpunkt: 136…138 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00462409 CAS-Nummer: 2364-75-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...