5-Chlor-2-nitropyridin 97+%
Lieferant: Apollo Scientific
|
Achtung
|
OR2032-25G
OR2032-5G
APOSOR2032-25GEA
29
EUR
APOSOR2032-25G
APOSOR2032-5G
5-Chlor-2-nitropyridin 97+%
5-Chlor-2-nitropyridin
Formel:
C₅H₃ClN₂O₂ Molecular Weight: 158.54 g/mol Schmelzpunkt: 120…123 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03092916 CAS-Nummer: 52092-47-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...