3-Chloro-4-methylphenylhydrazine hydrochloride

Lieferant: Apollo Scientific
Achtung

Synonyme: 3-Chloro-4-methylphenylhydrazine Hydrochloride

OR2034-100G
APOSOR2034-100GEA 195 EUR
APOSOR2034-100G
3-Chloro-4-methylphenylhydrazine hydrochloride
3-Chloro-4-methylphenylhydrazine hydrochloride
Formel: C₇H₉ClN₂·HCl
Molecular Weight: 193,08 g/mol
Schmelzpunkt: 230…231 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00012936
CAS-Nummer: 54812-56-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR