4-Hydroxy-3-nitropyridin-N-oxid 97+%

Lieferant: Apollo Scientific
Achtung

Synonyme: 4-Hydroxy-3-nitropyridine 1-oxide

OR2037-1G OR2037-5G
APOSOR2037-1GEA 34.8 EUR
APOSOR2037-1G APOSOR2037-5G
4-Hydroxy-3-nitropyridin-N-oxid 97+%
4-Hydroxy-3-nitropyridin-N-oxid
Formel: C₅H₄N₂O₄
Molecular Weight: 156,0963 g/mol
Schmelzpunkt: 225…226 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00234062
CAS-Nummer: 31872-57-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR