3-(Chloromethyl)-1-methyl-1H-pyrazole 97%

Lieferant: Apollo Scientific
Gefahr

OR2055-250MG OR2055-1G
APOSOR2055-250MGEA 109 EUR
APOSOR2055-250MG APOSOR2055-1G
3-(Chloromethyl)-1-methyl-1H-pyrazole 97%
3-(Chloromethyl)-1-methyl-1H-pyrazole
Formel: C₅H₇ClN₂
Molecular Weight: 130.58 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD08690275
CAS-Nummer: 84547-64-8
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR