1,2,3-Benzothiadiazole-5-carbaldehyde 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR2062-1G
OR2062-250MG
OR2062-500MG
APOSOR2062-1GEA
306
EUR
APOSOR2062-1G
APOSOR2062-250MG
APOSOR2062-500MG
1,2,3-Benzothiadiazole-5-carbaldehyde 95%
1,2,3-Benzothiadiazole-5-carbaldehyde
Formel:
C₇H₄N₂OS Molecular Weight: 164,19 g/mol Schmelzpunkt: 102…104 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD08690236 CAS-Nummer: 394223-15-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...