2-(Morpholinomethyl)phenylboronsäurepinakolester 95+%

Lieferant: Apollo Scientific
Gefahr

Synonyme: 4-(2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)morpholine

OR2071-1G OR2071-5G
APOSOR2071-1GEA 72.5 EUR
APOSOR2071-1G APOSOR2071-5G
2-(Morpholinomethyl)phenylboronsäurepinakolester 95+%
2-(Morpholinomethyl)phenylboronsäurepinakolester
Formel: C₁₇H₂₆BNO₃
Molecular Weight: 303.2043 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD07781208
CAS-Nummer: 876316-33-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR