2-Methylbenzothiazole-6-sulphonyl chloride 95+%
Lieferant: Apollo Scientific
|
Gefahr
|
OR2093-250MG
APOSOR2093-250MGEA
132
EUR
APOSOR2093-250MG
2-Methylbenzothiazole-6-sulphonyl chloride 95+%
2-Methylbenzothiazole-6-sulphonyl chloride
Formel:
C₈H₆ClNO₂S₂ Molecular Weight: 247,72 g/mol Schmelzpunkt: 98…101 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03990571 CAS-Nummer: 21431-13-0 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...