3-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure 96%

Lieferant: Apollo Scientific
Achtung

OR2096-1G OR2096-5G OR2096-25G
APOSOR2096-1GEA 55.1 EUR
APOSOR2096-1G APOSOR2096-5G APOSOR2096-25G
3-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure 96%
3-(2,3-Dimethylphenylcarbamoyl)phenylboronsäure
Formel: C₁₅H₁₆BNO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD09027262
CAS-Nummer: 957060-99-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR