3-(4-Bromophenyl)-1H-pyrazole

Lieferant: Apollo Scientific
Gefahr

OR21-10G OR21-5G OR21-25G
APOSOR21-10GEA 161 EUR
APOSOR21-10G APOSOR21-5G APOSOR21-25G
3-(4-Bromophenyl)-1H-pyrazole
3-(4-Bromophenyl)-1H-pyrazole
Formel: C₉H₇BrN₂
Molecular Weight: 223,08 g/mol
Schmelzpunkt: 132…136 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01940434
CAS-Nummer: 73387-46-9
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR